Learn More
Fisherbrand™ Natural Rubber One Hole Stoppers
Product Code. 11567612 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Bottom:
5 mm
7 mm
8 mm
9 mm
10 mm
11 mm
15 mm
17 mm
19 mm
21 mm
23 mm
25 mm
27 mm
29 mm
31 mm
33 mm
39 mm
43 mm
49 mm
57 mm
62 mm
67 mm
71 mm
75 mm
92 mm
105 mm
Diameter (Metric) Top:
7 mm
9 mm
10.5 mm
11.5 mm
12.5 mm
14 mm
18 mm
20.3 mm
22.5 mm
24.5 mm
26.5 mm
28.5 mm
31 mm
33 mm
36 mm
38.5 mm
44.5 mm
49 mm
55 mm
64 mm
69 mm
74 mm
78 mm
85 mm
102 mm
116.5 mm
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Pack of 50
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Bottom Diameter (Metric) Top unitSize
11567612 49 mm 55 mm Pack of 5
11577592 10 mm 12.5 mm Pack of 20
11587592 11 mm 14 mm Pack of 20
11517602 17 mm 20.3 mm Pack of 20
11527602 19 mm 22.5 mm Pack of 20
11507602 15 mm 18 mm Pack of 20
11567622 105 mm 116.5 mm Each
11547602 23 mm 26.5 mm Pack of 20
11537602 21 mm 24.5 mm Pack of 20
11557602 25 mm 28.5 mm Pack of 20
11567602 27 mm 31 mm Pack of 10
11577602 29 mm 33 mm Pack of 10
11587602 31 mm 36 mm Pack of 10
11547612 43 mm 49 mm Pack of 5
11597602 33 mm 38.5 mm Pack of 10
11527612 39 mm 44.5 mm Pack of 10
11587612 57 mm 64 mm Pack of 2
11527592 5 mm 7 mm Pack of 50
11597612 62 mm 69 mm Pack of 2
11507622 67 mm 74 mm Pack of 2
11517622 71 mm 78 mm Each
11527622 75 mm 85 mm Each
11547592 7 mm 9 mm Pack of 20
11557592 8 mm 10.5 mm Pack of 20
11547622 92 mm 102 mm Each
10410893 9 mm 11.5 mm Pack of 20
11567592 9 mm 11.5 mm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
Product Code. 11567612
Only null left
Add to basket
Add to basket
Used for laboratory purposes.
  • Made up of natural rubber
  • Red in color
  • Contains one hole
Specifications
Diameter (Metric) Hole 5mm
For Use With (Application) For laboratory use
Hardness 46/55 IRHD
Length (Metric) 42 mm
Quantity 5/Pk.
Color Red
Diameter (Metric) Bottom 49 mm
Diameter (Metric) Top 55 mm
Material Natural Rubber
Product Type Tapered Stopper (1 hole)