Learn More
Fisherbrand™ Natural HDPE Cylinder Bottles with White Polypropylene Caps
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
60 mL
120 mL
240 mL
480 mL
950 mL
Quantity:
12
24
48
77
180
240
800
Unit Size:
Pack of 12
Pack of 180
Pack of 24
Pack of 240
Pack of 48
Pack of 77
Pack of 800
Description
Natural HDPE Narrow-Mouth Cylinder Bottles:
- Shoulder style allows to pour small quantities without spilling
- Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping
Polypropylene PE Foam/PTFE Lined Caps:
- Offers good impact strength, durability, thermal resistance, and pliability
- Autoclavable
- Liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
- Liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference
Specifications
Specifications
| Capacity (Metric) | 60 mL |
| Capacity (English) | 2 oz. |
| Color | Natural |
| Material | HDPE |
| Quantity | 800 |
| Closure Color | White |
| Neck Finish | 24-410 |
| Closure Included | Yes |
| Mouth | Narrow |
| Product Type | Cylinder Bottle |
| Show More |