Learn More
RL Enterprises™ Natural Gas Micro Burner with Tétine Connector
Shop All Fisherbrand Products
Click to view available options
Height (Metric):
88 mm
Unit Size:
Each
Specifications
Specifications
| Gas Type | Natural Gas |
| Diameter (Metric) | 3 mm |
| For Use With (Application) | Tétine Connector |
| Height (Metric) | 88 mm |
| Includes | T-Tine Connector |
| No. per Pack | 1 Ea. |
| Weight (Metric) | 0.22 kg |