Description
- German, English, Spanish, French, Polish, Czech, Hungarian language settings
- SingleDISPLAY provides adaptive multifunctional digital PID-microprocessor controller with high-definition TFT-color display
- Digital backwards counter with target time setting, adjustable from 1 minute to 99 days
- Process time does not start until the set temperature is reached
- Three freely selectable temperature values
- Adjust temperature (Celsius or Fahrenheit), air flap position, program time, time zones, summertime/wintertime
Specifications
Specifications
| Certifications/Compliance | CE |
| Capacity (Metric) | 108 L |
| For Use With (Application) | Applied for sterilisation of medical materials, complete killing of highly resistant microorganisms, special application of depyrogenisation |
| Includes | Works calibration certificate |
| Model | SN110 |
| Width (Metric) | 745 mm |
| Width (Metric) Interior | 560 mm |
| Height (Metric) | 864 mm |
| Height (Metric) Interior | 480 mm |
| Depth (Metric) | 584 mm |
| Show More |