Learn More
Memmert™ Natural Convection Hot Air Sterilizer with SingleDISPLAY
Product Code. 12885843 Shop All Memmert Products
Change view
Click to view available options
Capacity (Metric):
32 L
53 L
74 L
108 L
161 L
256 L
449 L
749 L
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12885843 74 L Each
12895843 108 L Each
12805853 161 L Each
12815853 256 L Each
12875843 32 L Each
12825853 449 L Each
12865843 53 L Each
12835853 749 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 12885843 Supplier Memmert™ Supplier No. SN75
Only null left
Add to basket
Add to basket
Sterilize a variety of containers and instruments with this hot air sterilizer. Memmert™ Natural Convection Hot Air Sterilizers with SingleDISPLAY allow adjustment of pre-heated fresh air admixture by air flap control in 10% steps. Safer than disinfection.
  • German, English, Spanish, French, Polish, Czech, Hungarian language settings
  • SingleDISPLAY provides adaptive multifunctional digital PID-microprocessor controller with high-definition TFT-color display
  • Digital backwards counter with target time setting, adjustable from 1 minute to 99 days
  • Process time does not start until the set temperature is reached
  • Three freely selectable temperature values
  • Adjust temperature (Celsius or Fahrenheit), air flap position, program time, time zones, summertime/wintertime
Specifications
Certifications/Compliance CE, cUL, DVE, EAC
Capacity (Metric) 74 L
Type Natural Convection Hot Air Sterilizer
Display TFT
For Use With (Application) Applied for sterilisation of medical materials, complete killing of highly resistant microorganisms, special application of depyrogenisation
Includes Works Calibration Certificate
Material Stainless Steel
Model SN75
Dimensions (L x W x H) 585 x 944 x 514 mm
Length (Metric) 514 mm
Width (Metric) 585 mm
Width (Metric) Interior 400 mm
Height (Metric) 944 mm
Height (Metric) Interior 560 mm
Depth (Metric) 514 mm
Depth (Metric) Interior 330 mm
Shipping Weight (Metric) 66 kg
Electrical Requirements 115/230 V 50/60 Hz
Voltage 115/230 V
Wattage 1800/2500 W
Frequency 50/60 Hz