Learn More
Memmert™ Natural Circulation Oil Baths with Excellent Temperature Controller
Product Code. 10403503 Shop All Memmert Products
Change view
Click to view available options
Capacity (Metric):
22 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10403503 22 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10403503 Supplier Memmert™ Supplier No. ONE22B02682
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Capacity (Metric) 22 L
Temperature Range 5°C to 200°C
Type Oil Bath
Material Stainless Steel
Warranty 3 Years
Length (Metric) 578 mm
Width (Metric) 436 mm
Height (Metric) 296 mm
Shipping Weight (Metric) 20 kg
Electrical Requirements 230 V 50/60 Hz 200 W
Voltage 230 V
Wattage 2 w
Frequency 50/60 Hz