missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NADK Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | NADK |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17581396
|
Proteintech
29290-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17571396
|
Proteintech
29290-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NADK antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human NADK. This antibody recognizes human, mouse, rat antigen. The NADK antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NADK | |
| Polyclonal | |
| Rabbit | |
| 65220 | |
| Primary |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Mouse, Rat | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title