Learn More
NADK Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 29290-1-AP-20UL
 View more versions of this product
Product Code. 17581396
  • 1710.33 NOK / 20µL
Description
The NADK antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human NADK. This antibody recognizes human, mouse, rat antigen. The NADK antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
NADK
Polyclonal
Rabbit
Primary
Human, Mouse, Rat
Western Blot, ELISA
Unconjugated
20 μL
65220