Learn More
NADK Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29290-1-AP-20UL
This item is not returnable.
View return policy
Description
The NADK antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human NADK. This antibody recognizes human, mouse, rat antigen. The NADK antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NADK | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot, ELISA | |
| Unconjugated | |
| 20 μL | |
| 65220 | |
| IgG |