Learn More
NAD+, MedChemExpress
Product Code. 16400938
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
16400938 5 g 5g
16422179 1 g 1g
2 options
Product Code. 16400938
Brand: Medchem Express HYB0445
MedChemExpress NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage.
NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage.
Specifications
Chemical Name or Material NAD+
CAS 53-84-9
Color White
Purity Grade Notes Research
Molecular Formula C21H27N7O14P2
Quantity 5 g
Synonym β-DPN β-NAD β-Nicotinamide Adenine Dinucleotide
Solubility Information H2O : 41.67 mg/mL (62.81 mM; Need ultrasonic)
SMILES O[C@H]([C@@H]1O)[C@@H](O[C@@H]1COP(OP(OC[C@H]2O[C@@H]([N+]3=CC=CC(C(N)=O)=C3)[C@H](O)[C@@H]2O)([O-])=O)(O)=O)N4C(N=CN=C5N)=C5N=C4
Molecular Weight (g/mol) 663.43
Formula Weight 663.43
Percent Purity 98.24%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism