missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N6-Methyl-2'-deoxyadenosine, 99%

Product Code. p-7043007
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15426669

Brand: Thermo Scientific Chemicals J64961.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2002-35-9 | C11H15N5O3 | 265.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2002-35-9
Molecular Formula C11H15N5O3
Molecular Weight (g/mol) 265.27
MDL Number MFCD00055999
InChI Key DYSDOYRQWBDGQQ-XLPZGREQSA-N
PubChem CID 168948
ChEBI CHEBI:7417
SMILES CNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1

Specifications

Color White
Quantity 1 g
Synonym n6-methyl-2'-deoxyadenosine, n6-me-dado, n 6-me-dado, adenosine, 2'-deoxy-n-methyl, unii-48qzw2ir9h, 2r,3s,5r-2-hydroxymethyl-5-6-methylamino-9h-purin-9-yl tetrahydrofuran-3-ol, n 6-methyl-2'-deoxyadenosine, n6-methyldeoxyadenosine, 2'-deoxy-n6-methylado, 48qzw2ir9h
Solubility Information Soluble in water
IUPAC Name (2R,3S,5R)-2-(hydroxymethyl)-5-[6-(methylamino)purin-9-yl]oxolan-3-ol
Formula Weight 265.27
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N6-Methyl-2'-deoxyadenosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.