Learn More
N-Formylfluoxetine, TRC
Product Code. 30132879
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132879 250 mg 250mg
30134592 50 mg 50mg
30010726 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132879 Supplier Toronto Research Chemicals Supplier No. TRCF699005250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 199188-97-1
Molecular Formula C18 H18 F3 N O2
Molecular Weight (g/mol) 337.34
Synonym N-Formylfluoxetine, N-Methyl-N-[3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]formamide
IUPAC Name N-methyl-N-[3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]formamide
SMILES CN(CCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2)C=O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C18H18F3NO2/c1-22(13-23)12-11-17(14-5-3-2-4-6-14)24-16-9-7-15(8-10-16)18(19,20)21/h2-10,13,17H,11-12H2,1H3
Formula Weight 337.129
Percent Purity >95%
Chemical Name or Material N-Formylfluoxetine
Grade HPLC