CAS RN 199188-97-1
CAS RN 199188-97-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-Formylfluoxetine, TRC
CAS: 199188-97-1 Molecular Formula: C18 H18 F3 N O2 Molecular Weight (g/mol): 337.34 Synonym: N-Formylfluoxetine,N-Methyl-N-[3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]formamide IUPAC Name: N-methyl-N-[3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]formamide SMILES: CN(CCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2)C=O
2
N-Formylfluoxetine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.