Learn More
N-Formylfluoxetine, Mikromol™
Description
N-Formylfluoxetine
Specifications
Specifications
| Chemical Name or Material | N-Formylfluoxetine |
| CAS | 199188-97-1 |
| Analyte or Component Names | N-Formylfluoxetine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H18 F3 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-Formylfluoxetine |
| InChI Formula | InChI=1S/C18H18F3NO2/c1-22(13-23)12-11-17(14-5-3-2-4-6-14)24-16-9-7-15(8-10-16)18(19,20)21/h2-10,13,17H,11-12H2,1H3 |
| SMILES | CN(CCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2)C=O |
| IUPAC Name | N-methyl-N-[3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]formamide |
| Show More |