missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-S-(4-methoxybenzyl)-L-cysteine, 95%, Thermo Scientific™

Product Code. 15439498
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15439498

Brand: Thermo Scientific Alfa Aesar H63045.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141892-41-3
Molecular Formula C26H25NO5S
Molecular Weight (g/mol) 463.548
MDL Number MFCD00065635
InChI Key IWZGYHFOLFRYPK-DEOSSOPVSA-N
Synonym fmoc-cys pmeobzl-oh, fmoc-s-p-methoxybenzyl-l-cysteine, fmoc-cys mob-oh, n-fmoc-s-4-methoxybenzyl-l-cysteine, fmoc-s-4-methoxybenzyl-l-cysteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-methoxybenzyl thio propanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-s-4-methoxybenzyl-l-cysteine, fmoc-cys 4-meobzl-oh, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, pubchem19018
PubChem CID 11236605
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(4-methoxyphenyl)methylsulfanyl]propanoic acid
SMILES COC1=CC=C(C=C1)CSCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Color White
Quantity 25 g
Formula Weight 463.55
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-S-(4-methoxybenzyl)-L-cysteine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.