accessibility menu, dialog, popup

UserName

CAS RN 141892-41-3

H3COSOHONHOO

CAS RN 141892-41-3

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-{[(4-methoxyphenyl)methyl]sulfanyl}propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-{[(4-methoxyphenyl)methyl]sulfanyl}propanoic acid
Molecular Weight (g/mol): 463.55
Molecular Formula: C26H25NO5S
InChi Key: IWZGYHFOLFRYPK-UHFFFAOYNA-N
SMILES: COC1=CC=C(CSCC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)