Learn More
N-Boc-N-ethylglycine, 97%
Product Code. 15494787
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15494787
Brand: Thermo Scientific Alfa Aesar H32112.06
Request Bulk or Custom Format
CAS: 149794-10-5 | C9H17NO4 | 203.238 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 149794-10-5
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00672508
InChI Key SPBIXXXFDSLALC-UHFFFAOYSA-N
Synonym boc-n-ethylglycine, n-boc-n-ethyl-glycine, boc-n-ethyl glycine, n-tert-butoxycarbonyl-n-ethylglycine, tert-butoxycarbonyl ethyl amino acetic acid, n-boc-n-ethylglycine, ethyl n-boc-glycine, pubchem23123, 2-tert-butoxycarbonyl ethyl amino acetic acid, n-1,1-dimethylethoxy carbonyl-n-ethylglycine
PubChem CID 12991451
IUPAC Name 2-[ethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid
SMILES CCN(CC(=O)O)C(=O)OC(C)(C)C
Specifications
Melting Point 87°C to 88°C
Quantity 5 g
Formula Weight 203.24
Percent Purity 97%
Chemical Name or Material N-Boc-N-ethylglycine