accessibility menu, dialog, popup

UserName

CAS RN 149794-10-5

CH3NOOHOOH3CH3CCH3

CAS RN 149794-10-5

IUPAC Name: 2-{[(tert-butoxy)carbonyl](ethyl)amino}acetic acid
Synonyms: [(tert-butoxycarbonyl)(ethyl)amino]acetic acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: SPBIXXXFDSLALC-UHFFFAOYSA-N
SMILES: CCN(CC(O)=O)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)