Learn More
N-Boc-4-oxo-L-proline tert-butyl ester, 97%
Product Code. 11357159
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 11357159
Brand: Thermo Scientific Alfa Aesar H27375.03
Request Bulk or Custom Format
CAS: 166410-05-5 | C14H23NO5 | 285.34 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 166410-05-5
Molecular Formula C14H23NO5
Molecular Weight (g/mol) 285.34
MDL Number MFCD06797013
InChI Key MPNWQUWKRDADHK-JTQLQIEISA-N
Synonym n-boc-4-oxo-l-proline tert-butyl ester, s-di-tert-butyl 4-oxopyrrolidine-1,2-dicarboxylate, boc-4-oxo-l-proline tert-butyl ester, 1,2-di-tert-butyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, boc-4-oxo-l-prolinetert-butylester, di-tert-butyl s-4-oxopyrrolidine-1,2-dicarboxylate, di-tert-butyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butoxycarbonyl-4-oxo-l-proline tert-butyl ester, 1-tert-butyloxycarbonyl-4-oxo-l-proline tert-butyl ester, s-4-oxopyrrolidine-1,2-dicarboxylic acid di-tert-butyl ester
PubChem CID 11346670
IUPAC Name ditert-butyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)C1CC(=O)CN1C(=O)OC(C)(C)C
Specifications
Melting Point 64°C to 67°C
Quantity 1 g
Formula Weight 285.35
Percent Purity 97%
Chemical Name or Material N-Boc-4-oxo-L-proline tert-butyl ester