accessibility menu, dialog, popup

UserName

CAS RN 166410-05-5

CH3H3CCH3OOONOOH3CCH3H3C

CAS RN 166410-05-5

IUPAC Name: 1,2-di-tert-butyl 4-oxopyrrolidine-1,2-dicarboxylate
Synonyms: 1,2-di-tert-butyl 4-oxopyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 285.34
Molecular Formula: C14H23NO5
InChi Key: MPNWQUWKRDADHK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C1CC(=O)CN1C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results