Learn More
N-Boc-4-methylene-L-proline, 97%, Thermo Scientific™
Product Code. p-7030196
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463767 250 mg 250mg
11322899 1 g 1g
11332899 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15463767 Supplier Thermo Scientific Alfa Aesar Supplier No. H27746.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
As important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry. The fluorous prolins were synthesized from commercially available N-Boc-4-methylene- L-proline.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 84348-38-9
Molecular Formula C11H17NO4
Molecular Weight (g/mol) 227.26
MDL Number MFCD01861787
InChI Key ULLGRIBXGPATMA-UHFFFAOYNA-N
Synonym n-boc-4-methylene-l-proline, s-1-tert-butoxycarbonyl-4-methylenepyrrolidine-2-carboxylic acid, 1-boc-s-4-methylenepyrrolidine-2-carboxylic acid, s-4-methylene-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 2s-1-tert-butoxycarbonyl-4-methylidenepyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid, 4-methylene-, 1-1,1-dimethylethyl ester, 2s, n-boc-4-methylene-l-prolne, n-t-boc-4-methylene-l-proline, 1-tert-butoxycarbonyl-4-methyleneproline
PubChem CID 21727417
IUPAC Name (2S)-4-methylidene-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(=C)CC1C(O)=O
Specifications
Melting Point 110°C to 114°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 227.26
Percent Purity 97%
Chemical Name or Material N-Boc-4-methylene-L-proline