accessibility menu, dialog, popup

UserName

CAS RN 84348-38-9

IUPAC Name: 1-[(tert-butoxy)carbonyl]-4-methylidenepyrrolidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-4-methylidenepyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 227.26
Molecular Formula: C11H17NO4
InChi Key: ULLGRIBXGPATMA-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(=C)CC1C(O)=O
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results