missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-methylene-L-proline, 97%, Thermo Scientific™

Product Code. 11332899
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332899 5 g 5g
15463767 250 mg 250mg
11322899 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11332899 Supplier Thermo Scientific Alfa Aesar Supplier No. H27746.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry. The fluorous prolins were synthesized from commercially available N-Boc-4-methylene- L-proline.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84348-38-9
Molecular Formula C11H17NO4
Molecular Weight (g/mol) 227.26
MDL Number MFCD01861787
InChI Key ULLGRIBXGPATMA-UHFFFAOYNA-N
Synonym n-boc-4-methylene-l-proline, s-1-tert-butoxycarbonyl-4-methylenepyrrolidine-2-carboxylic acid, 1-boc-s-4-methylenepyrrolidine-2-carboxylic acid, s-4-methylene-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 2s-1-tert-butoxycarbonyl-4-methylidenepyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid, 4-methylene-, 1-1,1-dimethylethyl ester, 2s, n-boc-4-methylene-l-prolne, n-t-boc-4-methylene-l-proline, 1-tert-butoxycarbonyl-4-methyleneproline
PubChem CID 21727417
IUPAC Name (2S)-4-methylidene-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(=C)CC1C(O)=O

Specifications

Melting Point 110°C to 114°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 227.26
Percent Purity 97%
Chemical Name or Material N-Boc-4-methylene-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.