Learn More
Thermo Scientific™ N-Acetyl-Val-Ala-Asp-al
For cellular and molecular biology applications
668.00 NOK - 2077.00 NOK
Specifications
Purification Method | HPLC |
---|---|
Content And Storage | -30°C to -10°C |
Format | Lyophilized Powder |
Molecular Weight (g/mol) | 329.35 |
Product Type | N-Acetyl-Val-Ala-Asp-al |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488149
|
Thermo Scientific™
J66696.M^ |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15478149
|
Thermo Scientific™
J66696.#0 |
5 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
HPLC | |
Lyophilized Powder | |
N-Acetyl-Val-Ala-Asp-al | |
Acetyl-VAD-CHO |
-30°C to -10°C | |
329.35 | |
RUO - research use only |
For Research Use Only