missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-Homocysteine Thiolactone, MP Biomedicals™

Product Code. 11358399 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358399 100 g 100g
11471585 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358399 Supplier MP Biomedicals Supplier No. 0210009780

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

N-Acetyl-DL-Homocysteine Thiolactone, MP Biomedicals is a reagent for insolubilizing antibodies.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17896-21-8
Molecular Formula C6H9NO2S
Molecular Weight (g/mol) 159.20
MDL Number MFCD00005480
InChI Key NRFJZTXWLKPZAV-UHFFFAOYNA-N
Synonym citiolone, citiolase, thioxidrene, ahctl, achtl, ahct, dl-n-acetylhomocysteine thiolactone, n-acetylhomocysteine thiolactone, acetamide, n-tetrahydro-2-oxo-3-thienyl, 3-acetamidotetrahydro-2-thiophenone
PubChem CID 14520
IUPAC Name N-(2-oxothiolan-3-yl)acetamide
SMILES CC(=O)NC1CCSC1=O

Specifications

Melting Point 109°C to 111°C (literature)
Density 1.26g/cm3
Boiling Point 427.3°C (760 mmHg)
Refractive Index 1.54 (Lit.)
Quantity 100 g
Solubility Information Soluble in water (50mg/mL)
Formula Weight 159.203
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.