accessibility menu, dialog, popup

UserName

CAS RN 17896-21-8

CH3ONHSO

CAS RN 17896-21-8

IUPAC Name: N-(2-oxothiolan-3-yl)acetamide
Synonyms: citiolone
Molecular Weight (g/mol): 159.2
Molecular Formula: C6H9NO2S
InChi Key: NRFJZTXWLKPZAV-UHFFFAOYNA-N
SMILES: CC(=O)NC1CCSC1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results