Learn More
Macherey-Nagel™ N 20 Magnetic Crimp Caps
Brand: Macherey-Nagel™ 702928.9
Additional Details : Weight : 0.01000kg
Specifications
8 mm | |
Butyl Rubber, PTFE | |
3 mm |
Silver | |
Butyl Dark Gray/PTFE Gray |