Learn More
Macherey-Nagel™ N 20 Magnetic Crimp Caps
Shop All Macherey-Nagel Products
Click to view available options
Septum:
Butyl Red/PTFE Gray
Butyl/PTFE
Silicone/PTFE
Color (Seal):
Dark Gray/Gray
Light Gray/Dark Gray
Transparent Blue/Clear
Unit Size:
Pack of 100
Specifications
Specifications
| Color (Seal) | Dark Gray/Gray |
| Diameter (Metric) Opening | 8 mm |
| Color | Silver |
| Material | Butyl Rubber, PTFE |
| Septum | Butyl/PTFE |
| Thickness (Metric) | 3 mm |