Learn More
Macherey-Nagel™ N 20 Magnetic Crimp Caps
Brand: Macherey-Nagel™ 702929
Additional Details : Weight : 0.50000kg
Specifications
8 mm | |
PTFE, Silicone | |
100 Pk. |
Silver | |
Silicone Blue Transparent/PTFE Colorless | |
3 mm |