Learn More
Abnova™ MYBPHL (Human) Recombinant Protein
Human MYBPHL full-length ORF (ADR83131.1, 1 a.a. - 354 a.a.) recombinant protein with GST tag at N-terminal.
Brand: Abnova™ H00343263-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MEAATAPEVAAGSKLKVKEASPADAEPPQASPGQGAGSPTPQLLPPIEEHPKIWLPRALRQTYIRKVGDTVNLLIPFQGKPKPQAIWTHDGCALDTRRVSVRNGEQDSILFIREAQRADSGRYQLRVQLGGLEATATIDILVIERPGPPQSIKLVDVWGFSATLEWTPPQDTGNTALLGYTVQKADTKSGLWFTVLEHYHRTSCIVSDLIIGNSYAFRVFAENQCGLSETAPITTDLAHIQKAATVYKTKGFAQRDFSEAPKFTQPLANCTTVTGYNTQLFCCVRASPRPKIIWLKNKMDIQGNPKYRALTHLGICSLEIRKPGPFDGGIYTCKAVNPLGEASVDCRVDVKVPN
Specifications
ADR83131.1 | |
myosin binding protein H-like | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
343263 | |
Wheat germ expression system | |
10 μg | |
MYBPHL | |
GST |
Safety and Handling
- MYBPHL (Human) Recombinant Protein (P01)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : tris-HCl, reduced glutathione