missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MYBPHL (Human) Recombinant Protein
Human MYBPHL full-length ORF (ADR83131.1, 1 a.a. - 354 a.a.) recombinant protein with GST tag at N-terminal.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | ADR83131.1 |
---|---|
Gene ID (Entrez) | 343263 |
Name | myosin binding protein H-like |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16140244
|
Abnova™
H00343263-P01.10UG |
10 μg |
4115.00 NOK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16150244
|
Abnova™
H00343263-P01.25UG |
25 μg |
6245.00 NOK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MEAATAPEVAAGSKLKVKEASPADAEPPQASPGQGAGSPTPQLLPPIEEHPKIWLPRALRQTYIRKVGDTVNLLIPFQGKPKPQAIWTHDGCALDTRRVSVRNGEQDSILFIREAQRADSGRYQLRVQLGGLEATATIDILVIERPGPPQSIKLVDVWGFSATLEWTPPQDTGNTALLGYTVQKADTKSGLWFTVLEHYHRTSCIVSDLIIGNSYAFRVFAENQCGLSETAPITTDLAHIQKAATVYKTKGFAQRDFSEAPKFTQPLANCTTVTGYNTQLFCCVRASPRPKIIWLKNKMDIQGNPKYRALTHLGICSLEIRKPGPFDGGIYTCKAVNPLGEASVDCRVDVKVPN
Specifications
ADR83131.1 | |
myosin binding protein H-like | |
125% SDS-PAGE Stained with Coomassie Blue | |
MYBPHL | |
GST |
343263 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |