Learn More
Fisherbrand™ Sub-Gel Midi Horizontal Gel Electrophoresis Unit
Product Code. 11853303 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W x H):
220 x 125 x 90 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
11853303 220 x 125 x 90 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11853303
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
For rapid electrophoresis and quick checking of samples from PCR and cloning.
  • Runs up to 100 samples
  • Low buffer volumes
  • Injection-moulded construction
Specifications
Type Sub-Gel Midi Horizontal Gel Electrophoresis Unit
Gel Dimensions 70L x 100 mm W, 100L x 100 mm W
Sample Size 100
Volume (Metric) Buffer 300 mL
Dimensions (L x W x H) 220 x 125 x 90 mm