Learn More
Eppendorf™ Wall Mounting Device
Product Code. 10116081 Shop All Eppendorf Products
Click to view available options
Type:
Wall Mounting Device
Unit Size:
Each
Product Code. 10116081
Brand: Eppendorf™ 4981203009
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use this wall mount with Eppendorf pipette carousels. Eppendorf™ Wall Mounting Device is designed to hold the Varipette™ 4720.
  • Wall mount with adhesive surface for Varipette™ 4720
  • Pipette carousel holds up to 6 Eppendorf pipettes and/or other manual liquid handling instruments
TRUSTED_SUSTAINABILITY
Specifications
Type Wall Mounting Device
For Use With (Equipment) Varipette™ 4720
Quantity 1 Each