missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTRF1 Monoclonal Antibody (OTI5D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
2832.96 NOK - 6331.68 NOK
Specifications
| Antigen | MTRF1 |
|---|---|
| Clone | OTI5D6 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17567824
|
Origene Technologies
TA813119S |
30 μL |
2832.96 NOK
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17547824
|
Origene Technologies
TA813119 |
100 μL |
6331.68 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTRF1 Monoclonal antibody specifically detects MTRF1 in Human samples. It is validated for Western BlotSpecifications
| MTRF1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| A830062K05Rik; mitochondrial translational release factor 1; mitochontrial peptide chain release factor 1; MRF1; MRF-1; Mtrf1; MtRF-1; MTTRF1; Peptide chain release factor 1, mitochondrial; RF1 | |
| Human recombinant protein fragment corresponding to amino acids 240-445 of human MTRF1 (NP_004285) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| MTRF1 |
| OTI5D6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O75570 | |
| 9617 | |
| IgG2a | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title