missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTRF1 Monoclonal Antibody (OTI5D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813119
This item is not returnable.
View return policy
Description
MTRF1 Monoclonal antibody specifically detects MTRF1 in Human samples. It is validated for Western BlotSpecifications
| MTRF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75570 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 9617 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI5D6 | |
| Unconjugated | |
| MTRF1 | |
| A830062K05Rik; mitochondrial translational release factor 1; mitochontrial peptide chain release factor 1; MRF1; MRF-1; Mtrf1; MtRF-1; MTTRF1; Peptide chain release factor 1, mitochondrial; RF1 | |
| Human recombinant protein fragment corresponding to amino acids 240-445 of human MTRF1 (NP_004285) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction