missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH2 Monoclonal Antibody (OTI10F7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA814210S
Additional Details : Weight : 0.05000kg
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
MSH2 Monoclonal antibody specifically detects MSH2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MSH2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P43246, P43247, P54275 | |
Mouse | |
Protein A/G | |
RUO | |
17685, 4436, 81709 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
OTI10F7 | |
Unconjugated | |
Msh2 | |
AI788990; COCA1; DNA mismatch repair protein MSH2; FCC1; hMSH2; HNPCC; HNPCC1; LCFS2; mismatch repair ATPase MSH2; mismatch repair protein; MSH2; Msh2p; mutS homolog 2; mutS homolog 2 (E. coli); mutS homolog 2, colon cancer, nonpolyposis type 1; mutS protein homolog 2; O0935; PMS5; YOL090W | |
Human recombinant protein fragment corresponding to amino acids 1-304 of human MSH2 (NP_000242) produced in E.coli. | |
30 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |