Learn More
MSH2 Monoclonal Antibody (OTI10F7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
2816.72 NOK - 6295.38 NOK
Specifications
| Antigen | MSH2 |
|---|---|
| Clone | OTI10F7 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17508984
|
Origene Technologies
TA814210S |
30 μL |
2816.72 NOK
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17588974
|
Origene Technologies
TA814210 |
100 μL |
6295.38 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
MSH2 Monoclonal antibody specifically detects MSH2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| MSH2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| AI788990; COCA1; DNA mismatch repair protein MSH2; FCC1; hMSH2; HNPCC; HNPCC1; LCFS2; mismatch repair ATPase MSH2; mismatch repair protein; MSH2; Msh2p; mutS homolog 2; mutS homolog 2 (E. coli); mutS homolog 2, colon cancer, nonpolyposis type 1; mutS protein homolog 2; O0935; PMS5; YOL090W | |
| Human recombinant protein fragment corresponding to amino acids 1-304 of human MSH2 (NP_000242) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Msh2 |
| OTI10F7 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P43246, P43247, P54275 | |
| 17685, 4436, 81709 | |
| IgG1 | |
| Protein A/G | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.