missing translation for 'onlineSavingsMsg'
Learn More

MRPL24 Monoclonal Antibody (OTI1A10), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA813135

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 17547854

  • 4065.15 NOK / 100µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MRPL24 Monoclonal antibody specifically detects MRPL24 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

MRPL24
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q66H47, Q96A35, Q9CQ06
Mouse
Protein A/G
RUO
295224, 67707, 79590
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI1A10
Unconjugated
mrpl24
2010005E08Rik; 2810470K06Rik; 39S ribosomal protein L24, mitochondrial; 6720473G22Rik; AA407670; FLJ20917; hypothetical protein LOC532203; L24mt; MGC22737; MGC9831; Mitochondrial large ribosomal subunit protein uL24m; mitochondrial ribosomal protein L24; MRP-L18; MRPL24; MRP-L24; MRPL24 protein
Human recombinant protein fragment corresponding to amino acids 9-216 of human MRPL24 (NP_078816) produced in E.coli.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers