missing translation for 'onlineSavingsMsg'
Learn More

MRPL24 Monoclonal Antibody (OTI1A10), TrueMAB™, OriGene

Product Code. p-7225332
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
17567854 30 μL 30µL
17547854 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17567854

Brand: Origene Technologies TA813135S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRPL24 Monoclonal antibody specifically detects MRPL24 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen MRPL24
Applications Western Blot
Classification Monoclonal
Clone OTI1A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene mrpl24
Gene Accession No. Q66H47, Q96A35, Q9CQ06
Gene Alias 2010005E08Rik; 2810470K06Rik; 39S ribosomal protein L24, mitochondrial; 6720473G22Rik; AA407670; FLJ20917; hypothetical protein LOC532203; L24mt; MGC22737; MGC9831; Mitochondrial large ribosomal subunit protein uL24m; mitochondrial ribosomal protein L24; MRP-L18; MRPL24; MRP-L24; MRPL24 protein
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 9-216 of human MRPL24 (NP_078816) produced in E.coli.
Purification Method Protein A/G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295224, 67707, 79590
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.