missing translation for 'onlineSavingsMsg'
Learn More

MPEG-NH2 (Mn= 2,000), TRC

Product Code. 30467239
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
Product Code. Quantity unitSize
30467239 100 mg 100mg
30465197 50 mg 50mg
30467041 500 mg 500mg
3 options
This item is not returnable. View return policy

Product Code. 30467239

Brand: Toronto Research Chemicals TRCM745860100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80506-64-5
Molecular Formula (C2H4O)nC3H9NO
Molecular Weight (g/mol) 559.7

Specifications

Certifications/Compliance ISO 9001
Shipping Condition Room Temperature
Quantity 100 mg
Synonym MPEG-NH2 Amino∣Aminopoly(ethylene glycol) Methyl Ether∣Genamin MP 41-2000∣ME 200EA∣P 41-2000∣Monoamine-terminated Polyethylene Glycol Methyl Ether∣Monomethyl poly(ethylene glycol) Amine∣O-(2-Aminoethyl)-O-methoxypoly(ethylene glycol)∣O-(2-Aminoethyl)-O'-methylpolyethylene glycol∣PG 1AM2k∣Pluriol A 1020E Amine∣Pluriol A 2010E Amine∣Poly(ethylene Glycol) 2-Aminoethyl Methyl Ether∣Polyethylene Glycol Methyl 2-Aminoethyl Ether∣Polyethylene Glycol Mono(2-aminoethyl) Monomethyl Ether∣Sunbright ME 050EA∣Sunbright ME 100EA∣Sunbright ME 200EA∣Sunbright ME 400EA∣Sunbright ME 50EA∣mPEG 5000NH2∣alpha-Amino-ω-methoxy Polyoxyethylene∣alpha-Amino-ω-methoxypolyethylene Glycol∣alpha-Aminoethyl-ω-methoxy Polyethylene Glycol∣alpha-Methoxy-ω-aminopoly(ethylene Glycol)∣alpha-Methyl-ω-aminopoly(oxy-1,2-ethanediyl)∣alpha-Monomethoxy-ω-aminopoly(ethylene Glycol)∣
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.