Description
Defatted soybeans ground coarsely, used in animal research diets.
Specifications
Specifications
| Product Type | Animal Diet |
| Components | Crude Protein & gt;48%, Crude Fat & gt;0.5%, Crude Fiber <3.5% |
| Format | Powder |
| For Use With (Application) | Used in animal research diet |
| Particle Shape | Pellet |
| Quantity | 45.35 kg |
| Storage Requirements | 15°C to 30°C |