Learn More
MP Biomedicals Sodium Deficient Diet, Modified
Product Code. 11495672 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10,000 g
20,000 g
50,000 g
Unit Size:
10kg
20kg
50kg
Product Code. 11495672
Brand: MP Biomedicals undefined
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Sodium Deficient Diet
Components Casein Purified High Nitrogen 20%, Sucrose 67%, Alphacel, Non-nutritive Bulk 2%, Corn Oil 7%, Sodium Free Salt Mixture (Hartcroft and Eisenstein) 4% Plus MP Vitamin Diet Fortification Mixture
Format Pellet
Particle Shape Pellet
Quantity 20,000 g
Species Rat
Storage Requirements 2°C to 8°C