Learn More
MP Biomedicals Sodium Deficient Diet, Modified
Product Code. 11405682 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10,000 g
20,000 g
50,000 g
Unit Size:
10kg
20kg
50kg
Product Code. 11405682
Brand: MP Biomedicals 0290290250
Specifications
Product Type Sodium Deficient Diet
Components Casein Purified High Nitrogen 20%, Sucrose 67%, Alphacel, Non-nutritive Bulk 2%, Corn Oil 7%, Sodium Free Salt Mixture (Hartcroft and Eisenstein) 4% Plus MP Vitamin Diet Fortification Mixture
Format Pellet
Particle Shape Pellet
Quantity 50,000 g
Species Rat
Storage Requirements 2°C to 8°C