Learn More
MP Biomedicals Menhaden Oil Diet, High
Product Code. 11452941 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10,000 g
20,000 g
50,000 g
Unit Size:
10kg
20kg
50kg
Product Code. 11452941
Brand: MP Biomedicals 0296019520
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Menhaden Oil Diet
Components Casein Purified High Nitrogen 20%, DL-Methionine 0.3%, Sucrose 34.58%, Corn Starch 15%, Menhaden Oil 20%, Corn Oil 1%, Alphacel, Non-Nutritive Bulk 5%, DL-a-Tocopherol Powder (250 IU/g) 0.12%, AIN-76 Mineral Mix 4% Plus MP Vitamin Diet Fortification Mixture 1.2X Normal Amount.
Format Pellet
For Use With (Application) Feed for livestock and aquaculture, such as Salmon, Shrimp, Tilapia, Catfish, it is also used as a dietary supplement and as a raw material for products such as lipstick, it is also used for experimental Rodents
Particle Shape Powder
Quantity 20,000 g
Storage Requirements 15°C to 30°C