Learn More
MP Biomedicals™ Dextri-Maltose Supplement
Product Code. 11422801 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422801 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11422801 Supplier MP Biomedicals™ Supplier No. 0296004801
  • ONLINE EXCLUSIVE PRICE
    1185.00 NOK
    1127.05 NOK / 1kg
    Save 57.95 NOK
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Maltose and dextrins obtained by enzymatic digestion of corn flour by barley malt.
  • Maltodextrin, a polymer chain composed of D-glucose units linked by α (1,4) bonds
  • Used in the development of encapsulation and non-encapsulation vehicles for use in the food industry
  • Used as a protein Maillard-induced conjugation agent for improved solubility and foaming activity
  • As a sugar-matrix forming agent, maltodextrin is used in the development of orally disintegrating tablets (ODT)
  • Maltose and dextrins obtained by enzymatic digestion of corn flour by barley malt
Specifications
Product Type Animal Diet
Format Course Powder
For Use With (Application) Maltodextrin, a Polymer chain composed of D-glucose units linked by a (1, 4) bonds, is used in the development of encapsulation and nanencapsulation vehicles for use in the food industry, Maltodextrin is used as a protein Maillard-induced conjugation agent for improved solubility and foaming activity
Quantity 1 kg
Storage Requirements 15°C to 30°C