Learn More
MP Biomedicals™ AIN-76C Semipurified Diet, Pellet
Product Code. 11487231 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 kg
20 kg
50 kg
Unit Size:
10kg
20kg
50kg
Product Code. Quantity unitSize
11487231 10 kg 10kg
11497231 20 kg 20kg
11407241 50 kg 50kg
3 options
Product Code. 11487231
Brand: MP Biomedicals™ 0296029610
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rat or mouse, Pellets
  • AIN-76 diet with the amount of Vitamin K increased 10-fold (to 500 μg/Kg of diet)
  • 0.02% BHT (Butylated Hydroxy Toluene) per gram of corn oil.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Animal Diet
Components Casein Purified High Nitro gen 20%, DL-Methionine 0.3%, Sucrose 50%, Corn Starch 15%, Alphacel, Non-Nutritive Bulk 5%, Corn Oil 5%, Choline Bitartrate 0.2%, AIN Mineral Mix, AIN Vitamin Mix 3.5%
Format Pellet
For Use With (Application) Maintaining Mice and Rats colonies in the research lab
Quantity 10 kg
Species Rat, Mouse
Storage Requirements 15°C to 30°C