Learn More
- Home
- Products
- Immunoassay Reagents and Kits
- Singleplex and Multiplex Bead Based Immunoassays
- R&D Systems™ Mouse XL Cytokine Luminex™ Performance Premixed Kit
R&D Systems™ Mouse XL Cytokine Luminex™ Performance Premixed Kit
Product Code. 18792727
Change view
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
29 Plex
3 Plex
30 Plex
31 Plex
32 Plex
33 Plex
34 Plex
35 Plex
36 Plex
37 Plex
38 Plex
39 Plex
4 Plex
40 Plex
41 Plex
42 Plex
43 Plex
44 Plex
45 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Each
44 product options available for selection
| Product Code. | Quantity | Unit Size |
|---|---|---|
| 18792727 | 29 Plex | Each |
| 18482500 | 45 Plex | 1 set |
| 18431820 | 11 Plex | 1 set |
| 18412620 | 12 Plex | 1 set |
| 18790747 | 5 Plex | Each |
| 18730827 | 42 Plex | Each |
| 18791007 | 20 Plex | Each |
| 18781847 | 9 Plex | Each |
| 18712737 | 27 Plex | Each |
| 18753407 | 13 Plex | Each |
| 18791527 | 23 Plex | Each |
| 18713667 | 3 Plex | Each |
| 18712407 | 40 Plex | Each |
| 18730047 | 31 Plex | Each |
| 18754307 | 41 Plex | Each |
| 18781007 | 30 Plex | Each |
| 18742007 | 10 Plex | Each |
| 18723247 | 22 Plex | Each |
| 18780747 | 39 Plex | Each |
| 18741627 | 44 Plex | Each |
| 18744707 | 18 Plex | Each |
| 18763667 | 7 Plex | Each |
| 18744377 | 2 Plex | Each |
| 18743937 | 28 Plex | Each |
| 18771397 | 6 Plex | Each |
| 18700827 | 36 Plex | Each |
| 18712827 | 33 Plex | Each |
| 18790567 | 17 Plex | Each |
| 18783757 | 32 Plex | Each |
| 18701717 | 19 Plex | Each |
| 18753177 | 34 Plex | Each |
| 18761117 | 43 Plex | Each |
| 18750177 | 14 Plex | Each |
| 18730477 | 21 Plex | Each |
| 18764307 | 38 Plex | Each |
| 18754707 | 26 Plex | Each |
| 18712547 | 25 Plex | Each |
| 18764377 | 15 Plex | Each |
| 18764217 | 4 Plex | Each |
| 18774637 | 35 Plex | Each |
| 18733507 | 8 Plex | Each |
| 18722827 | 16 Plex | Each |
| 18763017 | 37 Plex | Each |
| 18750747 | 24 Plex | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 44 options available.
This item is not returnable.
View return policy
Product Code.
18792727
Supplier
R&D Systems™
Supplier No.
FCSTM2029
-
63255.00 NOK
59834.25 NOK / Pack of 1
Save 3420.75 NOK
5% Off
You must Sign In and have a business account to purchase this item.
Restricted Item
Estimated Shipment: 16-09-2026
Log in to see stock.
Log in to see stock.
Add to basket
Add to basket
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 29 Plex |
| Species | Mouse |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |