Learn More
R&D Systems™ Mouse VCAM-1/CD106 (AAA40545) VersaClone cDNA
Description
- Also known as: CD106; Vcam-1
- VCAM1 is a member of the Ig superfamily and is a cell surface sialoglycoprotein expressed by cytokine-activated endothelium.
- It is a type I membrane protein that mediates leukocyte-endothelial cell adhesion and signal transduction and may play a role in the development of artherosclerosis and rheumatoid arthritis.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mVcam1 |
| Accession Number | AAA40545 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2233 bp |
| Gene ID (Entrez) | 7412 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |