Learn More
BD Mouse TNF Enhanced Sensitivity Flex Set (Bead C8)
For measuring soluble analytes present in very low concentrations
Brand:  BD 562336
Additional Details : Weight : 0.04000kg
Product Code. 15881829
  • 4705.00 NOK / Each
Estimated Shipment: 24-05-2024
to see stock.
Description
Description
The BD CBA Mouse TNF Enhanced Sensitivity Flex Set is a bead-based immunoassay capable of measuring mouse Tumor Necrosis Factor (TNF) in serum and cell culture supernatant samples.
  • Species Reactivity:Mouse
Specifications
Specifications
TNF
1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Serum, Cell Culture Supernatant
Measures Mouse tumor necrosis factor, flow cytometry
Mouse
100 Tests
4°C
Mouse TNF enhanced sensitivity flex set (bead C8)