Learn More
BD Mouse TNF Enhanced Sensitivity Flex Set (Bead C8)
For measuring soluble analytes present in very low concentrations
Brand: BD 562336
Additional Details : Weight : 0.04000kg
Description
The BD CBA Mouse TNF Enhanced Sensitivity Flex Set is a bead-based immunoassay capable of measuring mouse Tumor Necrosis Factor (TNF) in serum and cell culture supernatant samples.
Mouse
Specifications
TNF | |
1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) | |
Serum, Cell Culture Supernatant | |
Measures Mouse tumor necrosis factor, flow cytometry |
Mouse | |
100 Tests | |
4°C | |
Mouse TNF enhanced sensitivity flex set (bead C8) |