Learn More
R&D Systems™ Mouse/Rat Osteopontin (OPN) Quantikine ELISA Kit
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
Description
The Mouse/Rat Osteopontin (OPN) Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat Osteopontin/OPN in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), urine (50 ul).
Specifications
Specifications
| Assay Range | 39.00 - 2500pg/mL |
| Assay Sensitivity | 8.5pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine |
| Specificity | Natural and Recombinant Mouse and Rat OPN. This assay cross-reacts 8.2% with Recombinant Human Osteopontin. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 20750 |
| Quantity | 1 x 96-well Plate |
| Sample Volume | Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (50uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |