missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Noggin ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Mouse Noggin quantitates mouse Noggin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Noggin.
Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.
Specifications
Specifications
| Accession Number | Nog; Noggin; Noggin precursor; SYM1; symphalangism 1 (proximal); SYNS1; SYNS1A |
| Assay Range | 0.23 to 50 ng/mL |
| Assay Sensitivity | 0.23 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 18121 |
| Gene Symbol | NOG |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction